Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGAComfort PAM™ Puncture Resistant™ Insole with Steel Plate

Personal anti-fatigue mat insole ergonomically designed by podiatrists, Dual layer 100% memory foam, Clinically proven field tested to reduce pain and fatigue, Max shock absorption, ASTM puncture resistant comfort

$32.95

Specifications

Color Black, Blue, Yellow
Material Memory Foam, Steel
Product Type Specs Puncture resistant insole with steel plate
Waterproof No
For Use With (Application) Construction and renovation environments, working mobile and outdoors or in industrial settings
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-813-517
View Documents
MEGAComfort
PAMPRW567
Women's: 5-7
Pair for $32.95
Add to cart
 
18-813-518
View Documents
MEGAComfort
PAMPRM67/W89
Men's: 6-7, Women's: 8-9
Pair for $32.95
Add to cart
 
18-813-519
View Documents
MEGAComfort
PAMPRM89/W1011
Men's: 8-9, Women's: 10-11
Pair for $32.95
Add to cart
 
18-813-520
View Documents
MEGAComfort
PAMPRM1011/W1213
Men's: 10-11, Women's: 12-13
Pair for $32.95
Add to cart
 
18-813-521
View Documents
MEGAComfort
PAMPRM1213
Men's: 12-13
Pair for $32.95
Add to cart
 
18-813-522
View Documents
MEGAComfort
PAMPRM1415
Men's: 14-15
Pair for $32.95
Add to cart
 
Description

Description

  • Personal Anti-Fatigue Mat (PAM™) puncture resistant insole
  • Uses patented combination of high density dual layer
  • 100% memory foam with encapsulated steel plate
  • Meets and exceeds ASTM F2413-11
  • Reduces pain and fatigue while increasing balance and comfort
  • Protects from sharp objects such as shards of glass, metal and nails
    • Effective in construction and renovation environments
  • Tested for penetration resistance and corrosion
  • For industrial (safety and construction), mobile and work outdoors
  • Silver ion anti-microbial technology in top cloth offering anti-bacterial, anti-perspiration and anti-odor
Specifications

Specifications

Black, Blue, Yellow
Puncture resistant insole with steel plate
Construction and renovation environments, working mobile and outdoors or in industrial settings
Meets ASTM F2423-11
Memory Foam, Steel
No
Boots
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit