Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Pam3CSK4, TLR1 and TLR2 Ligand
Applications: Functional, In vitro assay, In vivo assay, Ligand Activation
$167.50 - $265.00
Specifications
Concentration | 0.5 mg/mL |
---|---|
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Use With (Application) | Activation of TLR1 and TLR2 heterodimer. Stimulation of TLR1 and TLR2 has been reported with 10 to 100ng/mL. Use in vitro assay reported in scientific literature (PMID: 24105263) Use in functional reported in scientific literature (PMID 25511699). Use in In-vivo reported in scientific literature (PMID 25466255). Use in ligand activation reported in multiple pieces scientific literature. |
Target | Pam3CSK4, TLR1 and TLR2 Ligand |
Species | Human/Mouse |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP22529725
|
Novus Biologicals™
NBP2252970.025MG |
0.025 mg |
Each for $167.50
|
|
|||||
NBP225297
|
Novus Biologicals™
NBP225297 |
0.1 mg |
Each for $265.00
|
|
|||||
Description
Pam3CSK4, TLR1 and TLR2 Ligand is a synthetic tripalmitoylated lipohexapeptide analog of the immunologically active N-terminal portion of bacterial lipoprotein. It activates monocytes and macrophages and is also a potent activator of proinflammatory transcription factor NF-kB. Pam3CSK4, TLR1 and TLR2 Ligand is recognized by a heterodimer formed between TLR1 and TLR2. Pam3CSK4, TLR1 and TLR2 Ligand stimulation: If your cell line does not naturally express TLR1+ TLR2, co-transfect with plasmids pCMV/TLR1and pCMV/TLR2. Forty-eight hours after transfection, stimulate cells with 10 to 100ng/ml of Pam3CSK4 for 6 to 24 hours. Determine Pam3CSK4 stimulation using appropriate detection assays. Buffer: Sterile waterSpecifications
0.5 mg/mL | |
Activation of TLR1 and TLR2 heterodimer. Stimulation of TLR1 and TLR2 has been reported with 10 to 100ng/mL. Use in vitro assay reported in scientific literature (PMID: 24105263) Use in functional reported in scientific literature (PMID 25511699). Use in In-vivo reported in scientific literature (PMID 25466255). Use in ligand activation reported in multiple pieces scientific literature. | |
Human/Mouse |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Pam3CSK4, TLR1 and TLR2 Ligand |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title