Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
5g
Packaging:
Wide Mouth Amber Glass Bottle
85-85-8
C15H11N3O
249.273
RAXUMGMWXZYADR-OBGWFSINSA-N
1-2-pyridylazo-2-naphthol,unii-095b53y3xv,pan indicator,2-naphthalenol, 1-2-pyridinylazo,1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one,1-2-pyridylazo-2-hydroxynaphthalene,pan van,1-2-pyridyldiazenyl naphthalen-2-ol,1-pyridin-2-azo-2-naphthol,2-naphthalenol, 1-2-2-pyridinyl diazenyl
5376264
(1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3
85-85-8
C15H11N3O
249.273
RAXUMGMWXZYADR-OBGWFSINSA-N
1-2-pyridylazo-2-naphthol,unii-095b53y3xv,pan indicator,2-naphthalenol, 1-2-pyridinylazo,1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one,1-2-pyridylazo-2-hydroxynaphthalene,pan van,1-2-pyridyldiazenyl naphthalen-2-ol,1-pyridin-2-azo-2-naphthol,2-naphthalenol, 1-2-2-pyridinyl diazenyl
5376264
(1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3

High quality chemicals for laboratory and specialized industrial use

Quantity 5g
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 249.28
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material PAN