Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1g
5g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

85-85-8
C15H11N3O
249.273
RAXUMGMWXZYADR-OBGWFSINSA-N
5376264

High quality chemicals for laboratory and specialized industrial use

CAS 85-85-8
Molecular Formula C15H11N3O
Packaging Wide Mouth Amber Glass Bottle
InChI Key RAXUMGMWXZYADR-OBGWFSINSA-N
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3
IUPAC Name (1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
Molecular Weight (g/mol) 249.273
PubChem CID 5376264
Formula Weight 249.28
Quantity 1g
Chemical Name or Material PAN
Grade BAKER ANALYZED™ Reagent
Show More Show Less