Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan Dengue Virus (1-4) Mouse anti-Virus, Clone: 323, MilliporeSigma™

Mouse Monoclonal Antibody

$387.00 - $391.00

Specifications

Antigen pan Dengue Virus (1-4)
Clone 323
Applications ELISA, Flow Cytometry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2193100
View Documents
MilliporeSigma
MABF2193100UG
100 μg
Each for $387.00
Add to cart
 
MABF219325
View Documents
MilliporeSigma
MABF219325UG
25 μg
Each for $391.00
Add to cart
 
Description

Description

pan Dengue Virus (1-4) Monoclonal antibody specifically detects pan Dengue Virus (1-4) in Virus samples. It is validated for ELISA, Flow Cytometry.
Specifications

Specifications

pan Dengue Virus (1-4)
ELISA, Flow Cytometry
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at 2-8°C from date of receipt.
323
Unconjugated
Mouse
Virus
Purified recombinant Dengue virus NS1 protein expressed in mammalian cells.
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit