Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pan-Endothelium Marker, p Ser123, p Ser124 Antibody (HIS52) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200076
Description
Pan-Endothelium Marker Monoclonal specifically detects Pan-Endothelium Marker in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative).Specifications
Pan-Endothelium Marker (p Ser123, p Ser124, p ser123, p ser124) | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1-10 ug/ml, Western Blot (Negative) | |
Mouse | |
Affinity Purified | |
RUO | |
Rat | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
HIS52 | |
Unconjugated | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
The immunogen for this antibody is Pan endothelium. | |
0.1 mg | |
Primary | |
Store at 4C. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction