Learn More
PAN KRT Mouse anti-Human, Clone: PAN-CK, Abnova™
Mouse monoclonal antibody raised against human Pan KRT.
Manufacturer:  Abnova Corporation MAB12744
Catalog No. 89-934-724
Add to cart
Description
Description
Specifications
Specifications
PAN KRT
PAN-CK
Mouse monoclonal antibody raised against human Pan KRT.
In 10mM PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Mouse
IgG
250 uL
Store at 4°C. Do not freeze.
Monoclonal
Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Human Pan KRT
Protein A/G purification
RUO
Primary
Human