Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555984
Description
Immunogen sequence: MRSVNDIMPM IGARFYTQLD AAQMRNDVIE EDLAKEVQNG RLFRLLAKLG TINERPEFQK DPTWSETGDR YLLKLFRDHL FHQVTEAGAP WIDLSHIISC LNKLDAGVPE KISLISRDEK SVLVVTY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
PAN3 is functions in cytoplasmic mRNA decay. It is as part of the Pan nuclease complex, recruits polyadenylate-binding protein which in turn stimulates PAN2 nuclease activity.
Specifications
PAN3 | |
Polyclonal | |
Unconjugated | |
PAN3 | |
2700050F09Rik; A430027N15Rik; AU014670; PAB1P-dependent poly(A)-nuclease; PAB1P-dependent poly(A)-specific ribonuclease; PAB-dependent poly(A)-specific ribonuclease subunit 3; PAB-dependent poly(A)-specific ribonuclease subunit PAN3; Pabp1-dependent poly A-specific ribonuclease subunit Pan3; PABP-dependent poly(A) nuclease 3; PAN deadenylation complex subunit 3; PAN2-PAN3 deadenylation complex subunit Pan3; PAN3; PAN3 poly(A) specific ribonuclease subunit; PAN3 poly(A) specific ribonuclease subunit homolog; PAN3 polyA specific ribonuclease subunit homolog (S. cerevisiae); Poly(A)-nuclease deadenylation complex subunit 3; RP11-179F17.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
255967 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q58A45 | |
PAN3 | |
Recombinant protein corresponding to Human PAN3. Recombinant protein control fragment (Product #RP-90645). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction