Learn More
Alfa Aesar™ Pancreastatin (33-49), Porcine
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66504EXD
 View more versions of this product
Catalog No. AAJ66504EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1846.93
Porcine
Solid
RUO
Pancreastatin (33-49), porcine
HPLC
0.5mg
Gln-Glu-Glu-Glu-Glu-Glu-Thr-Ala-Gly-Ala-Pro-Gln-Gly-Leu-Phe-Arg-Gly-NH2
-30° to -10°C