Learn More
BioworldSupplier Diversity Partner Pancreatic Digest of Casein (PEP C), 500 g DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306200601
Pancreatic Digest of Casein (PEP C)General purpose growth peptone used for the cultivation of most bacteria and fungi and some protozoa Can be used for indole testing because of its high tryptophan content Is biologically free of carbohydrates and is recommended for fermentation studiesCan be used to determine nitrate reductionProvides nitrogen, vitamins, minerals, and amino acids in prepared culture mediaDehydrated Appearance: off-white to beige, homogeneous powderSpecifications:Total Nitrogen (TN): 11.5 – 14.2%Amino Nitrogen (AN): 3.6 – 4.9%Ash: ≤ 6.5%Moisture: ≤ 6.0%pH (2% Solution): 6.9 ± 0.2
Catalog No. 50-254-348