Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic Lipase Antibody (SR1851), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Pancreatic Lipase |
---|---|
Clone | SR1851 |
Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Pancreatic Lipase Monoclonal antibody specifically detects Pancreatic Lipase in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Pancreatic Lipase | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5406 | |
IgG | |
Affinity purified |
SR1851 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase | |
A synthesized peptide derived from human Pancreatic Lipase (Uniprot #: P16233) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title