Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic Polypeptide/PP Mouse anti-Human, DyLight 680, Clone: 1964, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243634FR

Catalog No. NBP243634FR


Add to cart

Description

Description

Pancreatic Polypeptide/PP Monoclonal antibody specifically detects Pancreatic Polypeptide/PP in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Pancreatic Polypeptide/PP
1964
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PPY
Recombinant protein encompassing a sequence within the center region of human PAF49. The exact sequence is proprietary.
Protein G purified
Stem Cell Markers
Primary
5539
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Obinepitide, pancreatic polypeptide (ppy), pancreatic polypeptide PNPPP, pancreatic polypeptide Y, pancreatic prohormone
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit