Learn More
Pancreatic Polypeptide/PP Mouse anti-Human, DyLight 755, Clone: 2G5D12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237432IR
Catalog No. NBP237432IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Pancreatic Polypeptide/PP
2G5D12
Western Blot, Flow Cytometry, ELISA
Purified
PPY
Purified recombinant fragment of human PPY (AA: 1-95) expressed in E. Coli.
Protein A or G purified
Stem Cell Markers
Primary
5539
Western Blot, Flow Cytometry, ELISA
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Obinepitide, pancreatic polypeptide (ppy), pancreatic polypeptide PNPPP, pancreatic polypeptide Y, pancreatic prohormone
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human