Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic trypsin inhibitor Antibody (18), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223588AF594
Description
Pancreatic trypsin inhibitor Monoclonal specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA.Specifications
Pancreatic trypsin inhibitor | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PTI | |
Aprotinin from bovine lung adsorbed onto aluminum hydroxide gel | |
0.1 mL | |
404172 | |
Bovine | |
IgG1 κ |
Western Blot, ELISA | |
18 | |
Western Blot, ELISA | |
Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor | |
Mouse | |
Protein A or G purified | |
Primary | |
Clone 18 binds bovine aprotinin immobilized on a solid phase or free in solution. | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction