Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic trypsin inhibitor Mouse anti-Bovine, Alexa Fluor™ 750, Clone: 18, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223588AF750

Catalog No. NP22388A70


Add to cart

Description

Description

Pancreatic trypsin inhibitor Monoclonal antibody specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Pancreatic trypsin inhibitor
18
Western Blot, ELISA
Conjugated, Purified
Mouse
IgG1 Kappa
0.1 mL
Primary
404172.0
Bovine
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor
Aprotinin from bovine lung adsorbed onto aluminum hydroxide gel
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Clone 18 binds bovine aprotinin immobilized on a solid phase or free in solution.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit