Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic trypsin inhibitor Mouse anti-Bovine, Clone: 21, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | Pancreatic trypsin inhibitor |
---|---|
Clone | 21 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP22358901
|
Novus Biologicals
NBP2235890.1ML |
0.1 mL |
Each for $436.00
|
|
NB019348
|
Novus Biologicals
NBP2235890.025ML |
0.025 mL |
Each for $176.85
|
|
Description
Pancreatic trypsin inhibitor Monoclonal specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA.Specifications
Pancreatic trypsin inhibitor | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
404172 | |
IgG1 κ | |
Protein A or G purified | |
Clone 21 binds bovine aprotinin immobilized on a solid phase or free in solution. |
21 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Bovine | |
Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor | |
Aprotinin from bovine lung adsorbed onto aluminum hydroxide gel | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title