Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pancreatin 1X

Extracted from porcine pancreas.

Manufacturer:  Thermo Scientific™ J1988028

Catalog No. AAJ1988028


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

  • Format: White to off-white powder stabilized with lactose
  • CAS: 8049-47-6
  • MDL Number: MFCD00131789
Specifications

Specifications

Pancreatin 1X
White to off-white
200g
Powder stabilized with lactose
Porcine pancreas
Powder stabilized with lactose
Safety and Handling

Safety and Handling

EINECSNumber : 232-468-9

Recommended Storage : Keep cold

Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit