Learn More
panCytokeratin Mouse anti-Human, Janelia Fluor 549, Clone: HEP 111, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250219JF549
Catalog No. NBP50219549
Add to cart
Description
Description
panCytokeratin Monoclonal antibody specifically detects panCytokeratin in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
panCytokeratin
HEP 111
Immunohistochemistry
Purified
KRT1
HEP2 epithelial cell line
Protein G purified
Cytoskeleton Markers, Protein Kinase, Signal Transduction
Primary
3848
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CK1, EHK, EHK1, EPPK, K1, Keratin 1, KRT1, KRT1A, NEPPK
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Human