Learn More
panCytokeratin Mouse anti-Human, Janelia Fluor 646, Clone: LDS103, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250458JF646
Catalog No. NBP50458646
Add to cart
Description
Description
panCytokeratin Monoclonal antibody specifically detects panCytokeratin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
panCytokeratin
LDS103
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
KRT1
Cytoskeletons of the human urothelial RT4 cell line.
Protein G purified
Cytoskeleton Markers, Protein Kinase, Signal Transduction
Primary
3848
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CK1, EHK, EHK1, EPPK, K1, Keratin 1, KRT1, KRT1A, NEPPK
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Human