Panduit™ Cable Tie Tools - Tool Controlled Tension and Cut-Off

Manufacturer: Panduit Corp Headquarter GS4H

 View more versions of this product

Catalog No. 19-888-036

  Check Availability
Add to Cart

    Specifications

    Product Type Cable Tie Tool
    Color Light Gray/Red
    For Use With (Application) Production, maintenance, or construction applications
    For Use With (Equipment) S, HS, LH, H cable ties
    Includes Durable powder coat finish
    Weight (English) 16 oz.
    Weight (Metric) 454g

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit