Learn More
Pan-Ty™ Miniature Cross-section Plenum-Rated Nylon 6.6 Cable Tie
Manufacturer:  Panduit Corp Headquarter PLT1MC
 View more versions of this product
Catalog No. 19-888-049
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CSA, CE, RoHS Compliant
3.9 in.
Nylon 6.6
0.098 in.
Natural
99mm
Cable Ties
2.5mm