Learn More
PANK2 Mouse anti-Canine, Human, Rat, Clone: OTI3H9, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA501321
Catalog No. 50-167-2467
Add to cart
Description
Description
Pantothenate kinase is an essential regulatory enzyme in CoA biosynthesis, catalyzing the cytosolic phosphorylation of pantothenate (vitamin B5), N-pantothenoylcysteine, and pantetheine. CoA is the major acyl carrier, playing a central role in intermediary and fatty acid metabolism. In both yeast and fly, each with only 1 pantothenate kinase gene, the null mutant is inviable. Mutations in PANK2 are the cause of pantothenate kinase-associated neurodegeneration (PKAN), formerly known as Hallervorden-Spatz syndrome (HSS). PKAN is an autosomal recessive neurodegenerative disorder associated with iron accumulation in the brain. Mutations in PANK2 are the cause of hypoprebetalipoproteinemia, acanthocytosis, retinitis pigmentosa, and pallidal degeneration (HARP).
Specifications
Specifications
PANK2
Monoclonal
Unconjugated
PANK2
Liquid
PANK2
Full length human recombit protein of human PANK2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3H9
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BZ23
C20orf48, HARP, HSS, NBIA1, PKAN
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
296167, 477168, 80025