Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PANK3 Mouse anti-Human, Clone: OTI3C4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525321

Catalog No. PIMA525321


Add to cart

Description

Description

This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is expressed most abundantly in the liver.
Specifications

Specifications

PANK3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H999
Pantothenate kinase 3; Pantothenic acid kinase 3; hPanK3;
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79646
Flow Cytometry, Western Blot
OTI3C4
Unconjugated
PANK3
Liquid
PANK3
Full length human recombit protein of PANK3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit