Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PANK4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15501220UL

 View more versions of this product

Catalog No. NBP1550220


Add to cart

Description

Description

PANK4 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

PANK4
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
PANK4
Synthetic peptides corresponding to PANK4(pantothenate kinase 4) The peptide sequence was selected from the N terminal of PANK4. Peptide sequence MAECGASGSGSSGDSLDKSITLPPDEIFRNLENAKRFAIDIGGSLTKLAY.
86 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:100-1:2000
Q9NVE7
DKFZp547M242, EC 2.7.1.33, FLJ10782, hPanK4, pantothenate kinase 4, Pantothenic acid kinase 4
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
55229
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit