Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pannexin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577358
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
The pannexin gene family encodes a second class of putative gap junction proteins and are highly conserved in invertebrates and mammals. Pannexins (Panx) are four-pass transmembrane proteins that oligomerize to form large pore ion and metabolite-permeable channels. Pannexin-1 (PANX1) and Pannexin-3 are closely related, while Pannexin-2 is a more distant relation. PANX1 is a transmembrane protein that forms a mechanosensitive ATP-permeable channel between adjacent cells and in the endoplasmic reticulum. PANX1 may play a role as a Ca2+ -leak channel to regulate ER Ca2+ homeostasis and regulates neural stem and progenitor cell proliferation.
Specifications
Pannexin 1 | |
Polyclonal | |
Unconjugated | |
PANX1 | |
Pannexin 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24145, 315435, 55991 | |
-20°C | |
Lyophilized |
Immunofluorescence, Immunohistochemistry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P60570, Q96RD7, Q9JIP4 | |
Panx1 | |
Peptide (C)KEPTEPKFKGLRLE, corresponding to amino acid residues 18- 31 of human Pannexin 1 | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction