Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pannexin 2 Polyclonal Antibody
Pannexin 2 Polyclonal Antibody
Supplier: Thermo Scientific PA5141115
This item is not returnable.
View return policy
Description
Pannexin 2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
Pannexin 2 | |
Polyclonal | |
Unconjugated | |
PANX2 | |
h PX2; hPANX2; pannexin 2; pannexin-2; PANX2; Px2; si:ch211-192n14.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362979, 406218, 56666 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P60571, Q6IMP4, Q96RD6 | |
PANX2 | |
Synthetic peptide within Human Pannexin 2 71-95. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction