Learn More
Motic™ Panthera E2 Compound LED Microscopes
Designed for the future, the Panthera E2 is ready for ambitious work in educational environments and small life science labs. Features new CCIS™ Plan Ultra Contrast Objectives with stunning image quality in Brightfield, Phase Contrast, Darkfield, and Polarization.
Supplier: Motic™ 1100104601117
This item is not returnable.
View return policy
Description
- Swiveling, 25° inclined Siedentopf head
- LED light indicator
- Coaxial coarse and fine focusing adjustment with tension adjustment
- Built-in, low position coaxial rackless stage with right hand control
- Focusable Abbe condenser with 0.9 numerical aperature
- Iris diaphragm and slot
- Diopter Adjustment: ±5
Specifications
10X | |
Reversed Quadruple | |
LED |
CCIS™ Plan UC Objectives (4X, 10X, 40X(S)) | |
Binocular | |
WF 10X/20 mm |