Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pantoprazole sodium salt hydrate, Acros Organics™

Click to view available options
Quantity:
1g
5g
718635-09-7
C16H14F2N3NaO4S·xH2O
405.35
CGJRLPRCWSHOFU-UHFFFAOYSA-N
pantoprazole sodium,pantoprazole sodium hydrate,pantozol hydrate,protonix hydrate,c16h14f2n3nao4s.h2o,sodium pantoprazole 1-hydrate,pantoprazole sodium hydrate hplc,5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methanesulfinyl-1-sodio-1,3-benzodiazole hydrate,5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl sulfinyl-1h-benzimidazole sodium salt hydrate
23684923
sodium;5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfinyl]benzimidazol-1-ide;hydrate
COC1=C(C(=NC=C1)CS(=O)C2=NC3=C([N-]2)C=CC(=C3)OC(F)F)OC.O.[Na+]
718635-09-7
C16H14F2N3NaO4S·xH2O
405.35
CGJRLPRCWSHOFU-UHFFFAOYSA-N
pantoprazole sodium,pantoprazole sodium hydrate,pantozol hydrate,protonix hydrate,c16h14f2n3nao4s.h2o,sodium pantoprazole 1-hydrate,pantoprazole sodium hydrate hplc,5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methanesulfinyl-1-sodio-1,3-benzodiazole hydrate,5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl sulfinyl-1h-benzimidazole sodium salt hydrate
23684923
sodium;5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfinyl]benzimidazol-1-ide;hydrate
COC1=C(C(=NC=C1)CS(=O)C2=NC3=C([N-]2)C=CC(=C3)OC(F)F)OC.O.[Na+]
Water 4 to 6%
Infrared Spectrum Authentic
Quantity 1g
Melting Point 151.0°C to 155.0°C
Solubility Information (1% in water) Clear solution
Formula Weight 405.35
Chemical Name or Material Pantoprazole sodium salt hydrate

GHS H Statement
Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning