Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Panx2, Mouse anti-Rat, ATTO680, Clone: S121A-31, Abnova™
Mouse Monoclonal Antibody
Manufacturer: Abnova Corporation MAB18388
Specifications
Panx2 | |
S121A-31 | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
- | |
Panx2 | |
Recombinant protein corresponding to amino acids 11-664 (all but first 10 amino acids, encompassing extracellular and cytoplasmic domains) of rat Panx2. | |
Monoclonal | |
100 μg | |
Store at -20°C. | |
Antibody |
Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Western Blot | |
ATTO680 | |
Liquid | |
pannexin 2 | |
362979 | |
Mouse | |
IgG1 | |
Primary | |
RUO | |
Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title