Learn More
Aviva Systems Biology Corp PAOX Antibody - middle region (ARP53848_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53848_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAOX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PAOX. Alternate names: DKFZp434J245, MGC45464, PAO, RP11-122K13.11ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61147