Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAPOLA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548259

Catalog No. PA548259

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

Polymerase that creates the 3 ft.-poly(A) tail of mRNA ft.s. Also required for the endoribonucleolytic cleavage reaction at some polyadenylation sites. May acquire specificity through interaction with a cleavage and polyadenylation specificity factor (CPSF) at its C-terminus.
Specifications

Specifications

PAPOLA
Polyclonal
PBS with 0.09% sodium azide
P51003
Poly(A) polymerase alpha, PAP-alpha, Polynucleotide adenylyltransferase alpha, PAPOLA, PAP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10914
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PAPOLA
Liquid
PAPOLA
KLH conjugated synthetic peptide between 407-435 amino acids from the Central region of human PAP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit