Learn More
MilliporeSigma™ Papanicolaou's solution 1b Hematoxylin solution S
For cytological cancer and cycle diagnosis
$64.03 - $242.14
Specifications
Packaging Plastic Bottle
Storage Requirements 15° to 25°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M1092540500 View Documents MilliporeSigma™
1.09254.0500
500mL Each for $64.03
Add to cart
M1092542500 View Documents MilliporeSigma™
1.09254.2500
Each for $242.14
Add to cart
Description
Description
  • Provides good nucleic staining in 2 to 3 min.
  • Nuclei are stained blue, dark violet to black
Specifications
Specifications
Plastic Bottle
15° to 25°C