Learn More
MilliporeSigma™ Papanicolaou's solution 2a Orange G solution (OG 6)
Aids in diagnosis of cytological cancer and cycle
$259.00
Specifications
Storage Requirements 15° to 25°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
M1068882500
View Documents
MilliporeSigma™
1.06888.2500
2.5L Plastic Bottle
Each for $259.00
Add to cart
M1068881000
View Documents
MilliporeSigma™
1.06888.1000
1L Plastic Bottle This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Standard method in cancer and hormone cycle diagnosis
  • Smears with keratinized cells under acidic pH conditions
  • Gives intense reddish staining result with mature and keratinized squamous cells
  • Gives gentle yellow-orange staining result with mature and keratinized squamous cells
Specifications
Specifications
15° to 25°C