Learn More
MilliporeSigma™ Papanicolaou's Solution 3b Polychromatic
Used for human-medical cell diagnosis
Supplier:  MilliporeSigma™ 1.09272.0500
 View more versions of this product
Catalog No. M1092720500
Add to Cart
Description
Description
  • Ready to use
  • Staining solution for cytological cancer cycle diagnosis
  • Used for cytological investigation of sample material of human origin
  • Used to demonstrate the differentiation of squamous cells
Specifications
Specifications
Plastic Bottle
500mL
15° to 25°C