Papanicolaou Stain, EA-36-50 Formula, Counterstain for use with Hematoxylin Stains in the Papanicolaou Staining Procedure, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 550032

Packaging Poly natural
Quantity 1L

 View more versions of this product

Catalog No. 5500-32

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64-17-5,7732-18-5,17372-87-1,12067-99-1,10114-58-6,554-13-2
    CAS Min % 90.0,6.0,0.2,0.23,0.06,9.0E-4
    CAS Max % 91.0,8.0,0.3,0.24,0.06,9.0E-4
    Physical Form Murky Liquid
    Chemical Name or Material Papanicolaou Stain, EA-36-50 Formula

    Specifications

    Physical Form Murky Liquid
    Color Green
    Quantity 1L
    CAS 554-13-2,17372-87-1,10114-58-6,64-17-5,12067-99-1,7732-18-5
    CAS Min % 0.23,9.0E-4,90.0,6.0,0.06,0.2
    CAS Max % 0.3,0.24,0.06,9.0E-4,91.0,8.0
    Packaging Poly natural
    Chemical Name or Material Papanicolaou Stain, EA-36-50 Formula

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit