Learn More
AdipoGenSupplier Diversity Partner PAPAVERINE HCL(25GR) DFS Item
Manufacturer:  AdipoGenSupplier Diversity Partner CDXP0164G025
Papaverine . hydrochloride, AdipoGen Life Sciences. Chemical. CAS: 61-25-6. Formula: C20H21NO4 . HCl. MW: 339.4 . 36.5. Synthetic. Alkaloid antispasmodic drug that most usually treats visceral spasm or vasospasm. It has also been used in the treatment of erectile dysfunction. Smooth muscle relaxant and vasodilator. Phosphodiesterase PDE10A inhibitor. Show to have antiviral and antipsychotic activity.
Catalog No. 50-114-7228