Alfa Aesar™ Papaverine hydrochloride, 99%

Manufacturer: Alfa Aesar™ B2541214

Quantity 25g

 View more versions of this product

Catalog No. AAB2541214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61-25-6
    Molecular Formula C20H21NO4·HCl
    Formula Weight 375.85
    MDL Number MFCD00012745
    Merck Index 14,7019
    Synonym 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxyisoquinoline hydrochloride
    Chemical Name or Material Papaverine hydrochloride

    Specifications

    Formula Weight 375.85
    TSCA TSCA
    Molecular Formula C20H21NO4·HCl
    MDL Number MFCD00012745
    Merck Index 14,7019
    Synonym 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxyisoquinoline hydrochloride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Melting Point ≈220°C (dec.)
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number NW8575000
    CAS 61-25-6
    Chemical Name or Material Papaverine hydrochloride
    Assay 99%

    Description

    Solubility
    Soluble in water (50 mg/ml), DMSO, methanol, 100% ethanol and chloroform.

    Notes
    Light sensitive. Incompatible with oxidizing agents.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit