Learn More
Brady™ Paper Cardstock Tag: DANGER CURRENT ON SIGNED BY: DATE:
Identify facility equipment to your exact specifications. Brady™ Paper Cardstock Tags are durable two-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 47344
Catalog No. 19-113-441
Add to cart
Description
Description
  • Heavy duty paper tags are made of a durable cardstock material
  • They provide an economical and convenient way to ID equipment or areas of your facility
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor use
Specifications
Specifications
Black/red on white
General equipment
Not laminated
Rectangle
Paper cardstock
Header: DANGER
3 in. x 5.75 in.
(100) 12 in cotton string ties
100/Pack
-40°F to 194°F
CURRENT ON SIGNED BY: DATE: