Learn More
Brady™ Paperboard Warning Sign: GASOLINE
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
Specifications
Shape | Rectangular |
---|---|
Displayed Text | GASOLINE |
Temperature Range (English) | -40°F to 175°F |
Finish | Not Laminated |
Material | Paper |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-104-854
|
Brady™
115958 |
14 x 10 in. | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
19-104-855
|
Brady™
115959 |
10 x 7 in. | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
- Paperboard material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- For indoor use
Specifications
Rectangular | |
-40°F to 175°F | |
Paper | |
Non-adhesive | |
Corner holes | |
-40°F |
GASOLINE | |
Not Laminated | |
1 Ea. | |
Paper board | |
175°F | |
0.14 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.