Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAPP-A Human ELISA Kit Non-distribution product as customer accommodation.

Sandwich ELISA Kit

$494.00

Specifications

Storage Requirements 2°C to 8°C
Assay Sensitivity 0.6 ng/mL
Assay Range 0.686-500 ng/mL
Test Time 1 hr. 20 min.
Sample Volume Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EHPAPPA Non-distribution product as customer accommodation.
View Documents
Invitrogen™
EHPAPPA
96 Tests
Pack for $494.00
Add to cart
 
Description

Description

The Human PAPP-A ELISA quantitates Hu PAPP-A in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu PAPP-A. Principle of the method The Human PAPP-A solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes a secreted metalloproteinase which cleaves insulin-like growth factor binding proteins (IGFBPs). It is thought to be involved in local proliferative processes such as wound healing and bone remodeling. Low plasma level of this protein has been suggested as a biochemical marker for pregnancies with aneuploid fetuses.
Specifications

Specifications

2°C to 8°C
0.686-500 ng/mL
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
<10%
0.6 ng/mL
1 hr. 20 min.
<12%
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit