Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Paprotrain, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Reversible, non-ATP competitive inhibitor of mitotic kinesin-like protein 2 (MKLP-2) (Ki = 3.4 μM). Inhibits the basal ATPase activity of MKLP-2 (IC50 = 1.35 μM). Exhibits selectivity for MKLP-2 over 12 other members of the kinesin superfamily, including the closely-related MKLP-1. Cell permeable.
Specifications
Quantity | 10 mg |
Formula Weight | 245.28 |
Percent Purity | >99% |
Chemical Name or Material | Paprotrain |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction