Learn More
PAPSS2 Mouse anti-Human, Clone: OTI7E8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF807112
Catalog No. 50-166-8124
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Sulfation is a common modification of endogenous (lipids, proteins, and carbohydrates) and exogenous (xenobiotics and drugs) compounds. In mammals, the sulfate source is 3 ft.-phosphoadenosine 5 ft.-phosphosulfate (PAPS), created from ATP and inorganic sulfate. Two different tissue isoforms encoded by different genes synthesize PAPS. This gene encodes one of the two PAPS synthetases. Defects in this gene cause the Pakistani type of spondyloepimetaphyseal dysplasia. Two alternatively spliced transcript variants that encode different isoforms have been described for this gene.
Specifications
Specifications
PAPSS2
Monoclonal
Unconjugated
PAPSS2
Lyophilized
PAPSS2
Human recombit protein fragment corresponding to amino acids 1-247 of human PAPSS2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI7E8
PBS with 8% trehalose and no preservative; pH 7.3
O95340
ATPSK2, BCYM4, SK2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9060