Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAQR9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562690
Description
Immunogen sequence: IMLESWLFDL RGENPTLFVH F Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Specifications
PAQR9 | |
Polyclonal | |
Unconjugated | |
PAQR9 | |
Membrane progesterone P4 receptor epsilon; Membrane progesterone receptor epsilon; Membrane progestin receptor epsilon; mPR epsilon; PAQR9; Progesterone and adipoQ receptor family member 9; Progestin and adipoQ receptor family member 9; progestin and adipoQ receptor family member IX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
344838 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZVX9 | |
PAQR9 | |
Recombinant protein corresponding to Human PAQR9. Recombinant protein control fragment (Product #RP-101587). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction