Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PAR1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF3855SP
This item is not returnable.
View return policy
Description
PAR1/Thrombin Receptor Polyclonal specifically detects PAR1/Thrombin Receptor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
PAR1/Thrombin Receptor | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, F2R, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, ThrR, TRGPC, TRproteinase-activated receptor 1 | |
Goat | |
Affinity Purified | |
RUO | |
2149 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
P25116 | |
F2R | |
E. coli-derived recombinant human PAR1 Arg27-Thr102, Ser375-Thr425 Accession # P25116 | |
25 μg | |
Primary | |
Detects human PAR1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction