Learn More
Hampton Research Parabar 10312
SDP
Manufacturer:  Hampton Research HR2862
Encompass
Parabar 10312 is a viscous cryoprotectant for both small and large molecule crystallography. It is supplied as 5ml in an amber glass vial with a glass applicator rod in screw cap lid.
Catalog No. NC1146865
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.