Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paraffin, liquid, pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
8012-95-1
C15H11ClO7
338.696
MFCD00131611
FFNDMZIBVDSQFI-UHFFFAOYSA-N
delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
68245
CHEBI:38701
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
C1=C(C=C(C(=C1O)O)O)C2=C(C=C3C(=CC(=CC3=[O+]2)O)O)O.[Cl-]
8012-95-1
C15H11ClO7
338.696
MFCD00131611
FFNDMZIBVDSQFI-UHFFFAOYSA-N
delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
68245
CHEBI:38701
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
C1=C(C=C(C(=C1O)O)O)C2=C(C=C3C(=CC(=CC3=[O+]2)O)O)O.[Cl-]
Flash Point 218°C
Viscosity 65-75 mm2/s (40°C)
Specific Gravity 0.88
Packaging Glass bottle
Melting Point -24.0°C
Infrared Spectrum Authentic
Quantity 1L
Color Colorless
Density 0.8800g/mL
Boiling Point 300.0°C
Merck Index 14, 7186
Solubility Information Solubility in water: insoluble
Physical Form Oily Liquid
Grade Pure
Chemical Name or Material Paraffin, liquid, Pure
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 232-384-2