Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paraffin Oil, BAKER™, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
8012-95-1
C15H11ClO7
338.696
FFNDMZIBVDSQFI-UHFFFAOYSA-N
delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
68245
CHEBI:38701
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
C1=C(C=C(C(=C1O)O)O)C2=C(C=C3C(=CC(=CC3=[O+]2)O)O)O.[Cl-]
8012-95-1
C15H11ClO7
338.696
FFNDMZIBVDSQFI-UHFFFAOYSA-N
delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
68245
CHEBI:38701
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
C1=C(C=C(C(=C1O)O)O)C2=C(C=C3C(=CC(=CC3=[O+]2)O)O)O.[Cl-]

A grade of chemicals for laboratory use

Flash Point 135.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500mL
Specific Gravity 1L = 0.88kg
Chemical Name or Material Paraffin Oil