Learn More
Biochain Institute IncSupplier Diversity Partner Multiple Tumor Tissue Array - 40 types of tumors. Most of them have duplicates or triplicates from different individual, 5 slides/PK DFS Item
Manufacturer:  Biochain Institute IncSupplier Diversity Partner Z7020082
BioChain's tissue arrays or tissue microarrays (TMAs) are a useful tool in biomarker research and drug target validation. It consists of paraffin blocks that are assembled in array fashion to allow multiplex histological analysis. BioChain's TMAs provide a valuable tool for analysis of protein, RNA, or DNA molecules in oncology, both the malignant and metastatic markers. TMAs can also be constructed with tissues from other diseases and species.
Catalog No. 50-181-921