Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Biochain Institute IncSupplier Diversity Partner Cervical Tumor Tissue Array - 48 cases of uninvolved and cancerous tissues of the cervix in duplicates, 5 slides/PK DFS Item
Manufacturer:  Biochain Institute IncSupplier Diversity Partner Z7020016
BioChain's tissue arrays or tissue microarrays (TMAs) are a useful tool in biomarker research and drug target validation. It consists of paraffin blocks that are assembled in array fashion to allow multiplex histological analysis. BioChain's TMAs provide a valuable tool for analysis of protein, RNA, or DNA molecules in oncology, both the malignant and metastatic markers. TMAs can also be constructed with tissues from other diseases and species.
Catalog No. 50-181-951