Learn More
Parainfluenza virus type 1 Mouse anti-Virus, Clone: HM226, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13627
Catalog No. 89-033-590
Add to cart
Description
Description
Mouse monoclonal antibody raised against parainfluenza virus type 1 (PIV-1).
  • High specificity
  • High reactivity
Specifications
Specifications
Parainfluenza virus type 1
HM226
Mouse monoclonal antibody raised against parainfluenza virus type 1 (PIV-1).
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with parainfluenza virus type 1 and 3 fusion protein.
ELISA, Immunofluorescence
Unconjugated
ELISA Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
IgG2a
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)